Please login to your online account to display your discounted pricing

Alfa Aesar™ (+/-)-1-Phenyl-1,2-ethanediol, 97%

$36.64 - $36.64

Chemical Identifiers

Chemical Name or Material (+/-)-1-Phenyl-1,2-ethanediol
CAS 93-56-1
Molecular Formula C8H10O2
Formula Weight 138.17
MDL Number MFCD00003546
Beilstein 1306723
Merck Index 14,8861
Synonym (+/-)-Styrene glycol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0455106

 
alfa aesar™
L0455106
5g Each for $36.64
Description & Specifications

Specifications

Formula Weight 138.17
CAS 93-56-1
TSCA TSCA
Assay 97%
Molecular Formula C8H10O2
MDL Number MFCD00003546
Beilstein 1306723
Synonym (+/-)-Styrene glycol
Melting Point 66° to 68°C
Flash Point 160°C (320°F)
RTECS Number KI2500000
Quantity 5g
Chemical Name or Material (+/-)-1-Phenyl-1,2-ethanediol
Merck Index 14,8861
Boiling Point 272° to 274°C