missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Phenyl-1,2,3,4-tetrahydroisoquinoline, TCI America™

Manufacturer:  TCI America P20561G

 View more versions of this product

Catalog No. P20561G



Specifications

Specifications

1-Phenyl-1,2,3,4-tetrahydroisoquinoline
100°C
22990-19-8
C15H15N
1,2,3,4-Tetrahydro-1-phenylisoquinoline
C1CNC(C2=CC=CC=C21)C3=CC=CC=C3
209.292
209.29
White
1g
≥98.0% (GC,T)
MFCD02179241
PRTRSEDVLBBFJZ-UHFFFAOYSA-N
1-phenyl-1,2,3,4-tetrahydroisoquinoline
100137
Crystal-Powder at 20°C

Chemical Identifiers

22990-19-8
209.292
PRTRSEDVLBBFJZ-UHFFFAOYSA-N
100137
C1CNC(C2=CC=CC=C21)C3=CC=CC=C3
C15H15N
MFCD02179241
1,2,3,4-Tetrahydro-1-phenylisoquinoline
1-phenyl-1,2,3,4-tetrahydroisoquinoline
Safety and Handling

Safety and Handling

TSCA : No

Documents