Learn More
1-Pentylamine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1167414
Description
1-Pentylamine is used as a solvent, as a raw material in the manufacture of a variety of other compounds, including dyes, emulsifiers, and pharmaceutical products and as a flavoring agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Pentylamine is used as a solvent, as a raw material in the manufacture of a variety of other compounds, including dyes, emulsifiers, and pharmaceutical products and as a flavoring agent.
Solubility
Soluble in alcohol, ether.
Notes
Air Sensitive. Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
1-Pentylamine | |
-55°C | |
103°C to 105°C | |
Amine-like | |
1.411 | |
25 g | |
505953 | |
14,598 | |
Soluble in alcohol,ether. | |
CCCCCN | |
87.166 | |
CHEBI:74848 | |
98% |
110-58-7 | |
0.751 | |
−1°C (30°F) | |
C5H13N | |
MFCD00008236 | |
UN1106 | |
Air Sensitive | |
amylamine, 1-aminopentane, pentylamine, 1-pentylamine, n-amylamine, 1-pentanamine, n-pentylamine, monoamylamine, norleucamine, amyl amine | |
DPBLXKKOBLCELK-UHFFFAOYSA-N | |
pentan-1-amine | |
8060 | |
87.17 |
Chemical Identifiers
110-58-7 | |
87.166 | |
DPBLXKKOBLCELK-UHFFFAOYSA-N | |
8060 | |
pentan-1-amine |
C5H13N | |
MFCD00008236 | |
amylamine, 1-aminopentane, pentylamine, 1-pentylamine, n-amylamine, 1-pentanamine, n-pentylamine, monoamylamine, norleucamine, amyl amine | |
CHEBI:74848 | |
CCCCCN |
Safety and Handling
GHS H Statement
H225-H331-H314-H318-H302-H312
Highly flammable liquid and vapor.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H225-H302+H312-H314-H331-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMYLAMINES
EINECSNumber : 203-780-2
RTECSNumber : SC0300000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only