missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Pentyl isothiocyanate, 96%

Manufacturer:  Alfa Aesar™ A1497214

 View more versions of this product

Catalog No. AAA1497214



Specifications

Specifications

1-Pentyl isothiocyanate
191°C to 193°C
629-12-9
MFCD00014444
1746691
SGHJUJBYMSVAJY-UHFFFAOYSA-N
1-isothiocyanatopentane
69415
96%
Pungent
25g
C6H11NS
UN2922
pentyl isothiocyanate, n-amyl isothiocyanate, 1-pentyl isothiocyanate, amyl isothiocyanate, n-pentyl isothiocyanate, pentylisothiocyanate, unii-kl6u8nl15r, n-amylisothiocyanate, 1-isothiocyanato-pentane, pentane,1-isothiocyanato
CCCCCN=C=S
129.221
129.23

Chemical Identifiers

629-12-9
129.221
SGHJUJBYMSVAJY-UHFFFAOYSA-N
69415
CCCCCN=C=S
C6H11NS
MFCD00014444
pentyl isothiocyanate, n-amyl isothiocyanate, 1-pentyl isothiocyanate, amyl isothiocyanate, n-pentyl isothiocyanate, pentylisothiocyanate, unii-kl6u8nl15r, n-amylisothiocyanate, 1-isothiocyanato-pentane, pentane,1-isothiocyanato
1-isothiocyanatopentane
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

Documents