Learn More
Alfa Aesar™ 1-Pentyl isothiocyanate, 96%
Manufacturer: Alfa Aesar™ A1497206
Specifications
1-Pentyl isothiocyanate | |
191°C to 193°C | |
629-12-9 | |
MFCD00014444 | |
1746691 | |
SGHJUJBYMSVAJY-UHFFFAOYSA-N | |
1-isothiocyanatopentane | |
69415 | |
96% |
Pungent | |
5g | |
C6H11NS | |
UN2922 | |
pentyl isothiocyanate, n-amyl isothiocyanate, 1-pentyl isothiocyanate, amyl isothiocyanate, n-pentyl isothiocyanate, pentylisothiocyanate, unii-kl6u8nl15r, n-amylisothiocyanate, 1-isothiocyanato-pentane, pentane,1-isothiocyanato | |
CCCCCN=C=S | |
129.221 | |
129.23 |
Chemical Identifiers
629-12-9 | |
129.221 | |
SGHJUJBYMSVAJY-UHFFFAOYSA-N | |
69415 | |
CCCCCN=C=S |
C6H11NS | |
MFCD00014444 | |
pentyl isothiocyanate, n-amyl isothiocyanate, 1-pentyl isothiocyanate, amyl isothiocyanate, n-pentyl isothiocyanate, pentylisothiocyanate, unii-kl6u8nl15r, n-amylisothiocyanate, 1-isothiocyanato-pentane, pentane,1-isothiocyanato | |
1-isothiocyanatopentane |
Safety and Handling
GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.