Learn More
1-Pentyl isocyanate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1339903
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1-Pentyl isocyanate | |
0.878 | |
32°C (89°F) | |
C6H11NO | |
CH3(CH2)4NCO | |
1 g | |
1746688 | |
pentyl isocyanate, pentylisocyanate, pentane, 1-isocyanato, 1-pentyl isocyanate, n-pentyl isocyanate, 1-isocyanato-pentane, amylisocyanat, n-pentylisocyanate, n-amyl isocyanate, acmc-1clhg | |
CCCCCN=C=O | |
113.16 | |
113.16 |
3954-13-0 | |
136°C to 137°C | |
Pungent | |
1.415 | |
MFCD00144846 | |
UN3080 | |
Moisture sensitive | |
VRVUKQWNRPNACD-UHFFFAOYSA-N | |
1-isocyanatopentane | |
206036 | |
97% |
Chemical Identifiers
3954-13-0 | |
113.16 | |
VRVUKQWNRPNACD-UHFFFAOYSA-N | |
206036 | |
CCCCCN=C=O |
C6H11NO | |
MFCD00144846 | |
pentyl isocyanate, pentylisocyanate, pentane, 1-isocyanato, 1-pentyl isocyanate, n-pentyl isocyanate, 1-isocyanato-pentane, amylisocyanat, n-pentylisocyanate, n-amyl isocyanate, acmc-1clhg | |
1-isocyanatopentane |
Safety and Handling
GHS H Statement
H301-H330-H334-H226-H315-H319-H335
Toxic if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P285-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P342+P311-P363-P370+P378q-P501c
H226-H302+H312+H332-H315-H318-H334-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ISOCYANATES, TOXIC, FLAMMABLE, N.O.S.
TSCA : No
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only