missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Pentenylboronic acid, 98%

$35.86 - $410.99

Chemical Identifiers

CAS 104376-24-1
Molecular Formula C5H11BO2
Formula Weight 113.95
MDL Number MFCD01074677
Beilstein 2038297
Chemical Name or Material 1-Pentenylboronic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1967706 SDS Alfa Aesar™
L1967706
5g Each for $410.99
AAL1967703 SDS Alfa Aesar™
L1967703
1g Each for $106.07
AAL19677MD SDS Alfa Aesar™
L19677MD
250mg Each for $35.86
Specifications Description & Specifications

Chemical Identifiers

104376-24-1
113.95
2038297
C5H11BO2
MFCD01074677
1-Pentenylboronic acid

Specifications

104376-24-1
C5H11BO2
113.95
2038297
98%
79° to 81°C
5g
MFCD01074677
1-Pentenylboronic acid

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning