missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Pentenylboronic acid, 97+%, ACROS Organics™

Chemical Identifiers

CAS 104376-24-1
CAS Min % 97.0
CAS Max % 100.0
Assay Percent Range 97+%
Molecular Formula C5 H11 B O2
Linear Formula CH3(CH2)2CH=CHB(OH)2
Formula Weight 113.95
MDL Number MFCD01074677
Chemical Name or Material 1-Pentenylboronic acid
Assay ≥97.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC365950010 SDS Thermo Fisher Scientific
AC365950010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

104376-24-1
100.0
C5 H11 B O2
113.95
1-Pentenylboronic acid
97.0
97+%
CH3(CH2)2CH=CHB(OH)2
MFCD01074677
≥97.0%

Specifications

113.95
White
97.0
C5 H11 B O2
97+%
Glass bottle
CH3(CH2)2CH=CHB(OH)2
≥97.0%
Shiny Small Flakes
1g
100.0
104376-24-1
MFCD01074677
Authentic
1-Pentenylboronic acid

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.