missing translation for 'onlineSavingsMsg'
Learn More

1-Pentene 98.0+%, TCI America™

Supplier:  TCI America P0316500ML

 View more versions of this product

Catalog No. P0316500ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1-Pentene
Colorless
C5H10
500 mL
1-pentene, pentene, propylethylene, pentylene, alpha-amylene, alpha-n-amylene, 1-pentylene, 1-n-pentene, unii-alp8m0lu81, pentene-1
CCCC=C
70.14
70.14
Liquid
109-67-1
30°C
MFCD00003567
1108
YWAKXRMUMFPDSH-UHFFFAOYSA-N
pent-1-ene
8004
≥98.0% (GC)

Chemical Identifiers

109-67-1
70.14
YWAKXRMUMFPDSH-UHFFFAOYSA-N
8004
CCCC=C
C5H10
MFCD00003567
1-pentene, pentene, propylethylene, pentylene, alpha-amylene, alpha-n-amylene, 1-pentylene, 1-n-pentene, unii-alp8m0lu81, pentene-1
pent-1-ene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0019

RTECSNumber : SB2179000

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions