missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Pentene, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC268660025

 View more versions of this product

Catalog No. AC268660025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

1-Pentene
−28°C
Glass bottle
0.64
30.0°C
-165.0°C
109-67-1
100.0
C5H10
MFCD00003567
15,7234
Solubility in water: 0.15g/L (20°C). Other solubilities: miscible with alcohol, ether and beznene, soluble in most organic solvents
CCCC=C
70.13
70.13
97%
0.6400g/mL
Authentic
1.3704 to 1.3724
0.202 mPa.s (20°C)
Colorless
2.5L
96.0
97%
CH3CH2CH2CH=CH2
01,21
1-n-pentene, 1-pentene, 1-pentylene, alpha-amylene, alpha-n-amylene, pentene, pentene-1, pentylene, propylethylene, unii-alp8m0lu81
YWAKXRMUMFPDSH-UHFFFAOYSA-N
pent-1-ene
8004
Liquid

Chemical Identifiers

109-67-1
70.13
YWAKXRMUMFPDSH-UHFFFAOYSA-N
8004
CCCC=C
C5H10
MFCD00003567
1-n-pentene, 1-pentene, 1-pentylene, alpha-amylene, alpha-n-amylene, pentene, pentene-1, pentylene, propylethylene, unii-alp8m0lu81
pent-1-ene
Safety and Handling

Safety and Handling

GHS H Statement
May be fatal if swallowed and enters airways.
Extremely flammable liquid and vapor.
Harmful to aquatic life with long lasting effects.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Ground/Bond container and receiving equipment.
Ta

GHS Signal Word: Danger

EINECSNumber : 203-694-5

SDS Documents