missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Penten-3-ol, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 130010050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1-Penten-3-ol | |
616-25-1 | |
100.0 | |
0.8390g/mL | |
25°C | |
96% min. (GC) | |
C5H10O | |
C2H5CH(OH)CH=CH2 | |
5 g | |
0.839 | |
Solubility in water: slightly soluble in water | |
CCC(O)C=C | |
86.13 | |
86.13 | |
Powder |
99% | |
97.5 | |
Yellow to Beige | |
114.0°C to 115.0°C | |
Authentic | |
Glass bottle | |
1.4220 to 1.4260 | |
MFCD00004573 | |
01,443 | |
1-penten-3-ol, ethyl vinyl carbinol, vinyl ethyl carbinol, 1-pentene-3-ol, 1-ethylallyl alcohol, alpha-ethylallyl alcohol, 1-penten-3-ol e, 1-penten-3-ol, r, fema no. 3584, ethylvinylcarbinol | |
VHVMXWZXFBOANQ-UHFFFAOYNA-N | |
pent-1-en-3-ol | |
12020 | |
97% |
Chemical Identifiers
616-25-1 | |
86.13 | |
VHVMXWZXFBOANQ-UHFFFAOYNA-N | |
12020 | |
CCC(O)C=C |
C5H10O | |
MFCD00004573 | |
1-penten-3-ol, ethyl vinyl carbinol, vinyl ethyl carbinol, 1-pentene-3-ol, 1-ethylallyl alcohol, alpha-ethylallyl alcohol, 1-penten-3-ol e, 1-penten-3-ol, r, fema no. 3584, ethylvinylcarbinol | |
pent-1-en-3-ol |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
GHS Signal Word: Warning
EINECSNumber : 210-472-1
TSCA : TSCA
RUO – Research Use Only