Please login to your online account to display your discounted pricing

1-Pentanesulfonic Acid, Sodium salt, Monohydrate, HPLC grade, EMD Millipore

$357.03 - $1,101.63

Chemical Identifiers

Chemical Name or Material 1-Pentanesulfonic Acid, Sodium salt, Monohydrate
Name Note LiChropur™, For Ion Pair Chromatography
CAS 22767-49-3
Physical Form Crystalline Solid
Molecular Formula C5H11NaO3S
Formula Weight 174.2
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MPX01012

 
emd millipore corporation
PX01012
25g Each for $357.03

MPX01013

 
emd millipore corporation
PX01013
100g Each for $1,101.63
Description & Specifications

Specifications

Physical Form Crystalline Solid
Formula Weight 174.2
Color White
Quantity 25g
Bulk Density 390kg/m3
Recommended Storage Store at +5°C to +30°C
Chemical Name or Material 1-Pentanesulfonic Acid, Sodium salt, Monohydrate
Name Note LiChropur™, For Ion Pair Chromatography
Molecular Formula C5H11NaO3S
CAS 22767-49-3
Safety and Handling

Store at +5°C to +30°C