Please login to your online account to display your discounted pricing

1-Pentanesulfonic Acid, Sodium salt, Monohydrate, HPLC grade, EMD Millipore


Quantity 100g

Manufacturer: emd millipore corporation  PX01013

 View more versions of this product

Catalog No. MPX01013

  • / Each
 

Description & Specifications

Chemical Identifiers

Color White
Formula Weight 174.2
Physical Form Crystalline Solid
Quantity 100g
Bulk Density 390kg/m3
Molecular Formula C5H11NaO3S
Chemical Name or Material 1-Pentanesulfonic Acid, Sodium salt, Monohydrate
Name Note LiChropur™, For Ion Pair Chromatography
CAS 22767-49-3
Recommended Storage Store at +5°C to +30°C

Specifications

Chemical Name or Material 1-Pentanesulfonic Acid, Sodium salt, Monohydrate
Name Note LiChropur™, For Ion Pair Chromatography
CAS 22767-49-3
Physical Form Crystalline Solid
Molecular Formula C5H11NaO3S
Formula Weight 174.2
Accessories

Store at +5°C to +30°C