missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Pentanesulfonic acid, sodium salt hydrate, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 270370250

 View more versions of this product

Catalog No. AC270370250



Specifications

Specifications

(10% aq. soln.) clear, colorless
Authentic
Glass bottle
>300.0°C
1266615-65-9
C5H11NaO3S·xH2O
MFCD00149548
n-Amylsulfonic acid, sodium salt
174.12
Crystalline Powder
1-Pentanesulfonic acid, sodium salt hydrate
9.6% max. (100°C, 3 hrs) (vacuum dried)
White
25g
99%
CH3(CH2)4SO3Na·xH2O
04, III, 23
(10% in water) Clear colorless
174.12
99%

Chemical Identifiers

1266615-65-9
174.12
n-Amylsulfonic acid, sodium salt
C5H11NaO3S·xH2O
MFCD00149548
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER

GHS Signal Word: Warning

SDS Documents