Please login to your online account to display your discounted pricing

1-Pentanesulfonic acid, sodium salt, 99%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC206110250

 View more versions of this product

Catalog No. AC206110250

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Crystals
Assay ≥98.0% (after ion exchange) (on dry substance) (Alkalimetry)
Formula Weight 174.2
Quantity 25g
Color White to pale yellow
Solubility Solubility in water: Soluble
Beilstein 04, III, 23
CAS Max % 100.0
Loss on Drying 2 % max. (1 g, 120°C)
Synonym n-Amylsulfonic acid, sodium salt
CAS Min % 98.5
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Chemical Name or Material 1-Pentanesulfonic acid, sodium salt
Packaging Glass bottle
Assay Percent Range 98+%
Molecular Formula C5H11O3SNa
Linear Formula CH3(CH2)4SO3Na
MDL Number MFCD00007541
CAS 22767-49-3

Specifications

Chemical Name or Material 1-Pentanesulfonic acid, sodium salt
CAS 22767-49-3
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.0% (after ion exchange) (on dry substance) (Alkalimetry)
Assay Percent Range 98+%
Molecular Formula C5H11O3SNa
Linear Formula CH3(CH2)4SO3Na
Formula Weight 174.2
MDL Number MFCD00007541
Synonym n-Amylsulfonic acid, sodium salt
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents