Learn More
1-Pentanesulfonic acid, sodium salt, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 206111000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1-Pentanesulfonic acid, sodium salt | |
98.5 | |
>300.0°C | |
2 % max. (1 g, 120°C) | |
98+% | |
C5H11NaO3S | |
MFCD00007541,MFCD00149548 | |
04, III, 23 | |
(10% in water at 25°C) Clear colorless | |
[Na+].CCCCCS([O-])(=O)=O | |
174.19 | |
174.2 | |
Crystals |
22767-49-3 | |
100.0 | |
White to Yellow | |
Authentic | |
Glass bottle | |
CH3(CH2)4SO3Na | |
100 g | |
sodium 1-pentanesulfonate, sodium pentane-1-sulfonate, sodium pentanesulfonate, 1-pentanesulfonic acid sodium salt, 1-pentanesulfonic acid, sodium salt, 1-pentylsodiumsulfonate, ipc-alks-5, 1-pentanesulfonate, sodium, pentanesulfonic acid na-salt, pentanesulfonic acidsodiumsalt | |
ROBLTDOHDSGGDT-UHFFFAOYSA-M | |
sodium;pentane-1-sulfonate | |
23664617 | |
98+% |
Chemical Identifiers
22767-49-3 | |
174.19 | |
ROBLTDOHDSGGDT-UHFFFAOYSA-M | |
23664617 | |
[Na+].CCCCCS([O-])(=O)=O |
C5H11NaO3S | |
MFCD00007541,MFCD00149548 | |
sodium 1-pentanesulfonate, sodium pentane-1-sulfonate, sodium pentanesulfonate, 1-pentanesulfonic acid sodium salt, 1-pentanesulfonic acid, sodium salt, 1-pentylsodiumsulfonate, ipc-alks-5, 1-pentanesulfonate, sodium, pentanesulfonic acid na-salt, pentanesulfonic acidsodiumsalt | |
sodium;pentane-1-sulfonate |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
GHS Signal Word: Warning
EINECSNumber : 245-208-4
RUO – Research Use Only