Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(Pentafluorophenyl)ethanol, 96%

$58.04 - $58.04

Chemical Identifiers

Chemical Name or Material 1-(Pentafluorophenyl)ethanol
CAS 830-50-2
Molecular Formula C8H5F5O
Formula Weight 212.12
MDL Number MFCD00004510
Beilstein 5268491
Synonym alpha-Methyl-2,3,4,5,6-pentafluorobenzyl alcohol; Methyl pentafluorophenyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0261009

 
alfa aesar™
L0261009
10g Each for $58.04
Description & Specifications

Specifications

Formula Weight 212.12
Chemical Name or Material 1-(Pentafluorophenyl)ethanol
Assay 96%
Melting Point 32° to 34°C
Molecular Formula C8H5F5O
MDL Number MFCD00004510
CAS 830-50-2
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Boiling Point 68° to 70°C (30mmHg)
Quantity 10g
Flash Point 87°C (188°F)
Beilstein 5268491
Synonym alpha-Methyl-2,3,4,5,6-pentafluorobenzyl alcohol; Methyl pentafluorophenyl carbinol
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.