Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Pentadecyne, 98%

$81.12 - $273.99

Chemical Identifiers

Chemical Name or Material 1-Pentadecyne
CAS 765-13-9
Molecular Formula CH3(CH2)12C≡CH
Formula Weight 208.39
MDL Number MFCD00015081
Beilstein 1751009
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0254004

  SDS
alfa aesar™
L0254004
2g Each for $81.12

AAL0254009

  SDS
alfa aesar™
L0254009
10g Each for $273.99
Description & Specifications

Specifications

Formula Weight 208.39
Chemical Name or Material 1-Pentadecyne
TSCA TSCA
Assay 98%
MDL Number MFCD00015081
CAS 765-13-9
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Boiling Point 129° to 131°C (10mmHg)
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula CH3(CH2)12C≡CH
Beilstein 1751009
Melting Point 10°C
Quantity 2g
Flash Point >90°C (194°F)

Applications
1-Pentadecyne is used in pharmaceutical research.

Solubility
Not miscible in water.

Notes
Store under inert gas. Air sensitive. Avoid excessive heat and light. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.