Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Pentadecanol, 99%

$64.80 - $64.80

Chemical Identifiers

Chemical Name or Material 1-Pentadecanol
CAS 629-76-5
Molecular Formula CH3(CH2)14OH
Formula Weight 228.42
MDL Number MFCD00004759
Beilstein 1700695
Synonym n-Pentadecyl alcohol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0367306

  SDS
alfa aesar™
L0367306
5g Each for $64.80
Description & Specifications

Specifications

Formula Weight 228.42
Boiling Point 269° to 271°C
MDL Number MFCD00004759
CAS 629-76-5
Beilstein 1700695
Melting Point 45° to 46°C
Chemical Name or Material 1-Pentadecanol
Assay 99%
TSCA TSCA
Quantity 5g
Synonym n-Pentadecyl alcohol
Molecular Formula CH3(CH2)14OH

Applications
1-Pentadecanol is employed as starting reagent for the asymmetric synthesis of jaspine. It acts as a fatty alcohol.

Solubility
Insoluble in water.

Notes
Incompatible materials are Strong oxidizing agents, Strong acids. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.