Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(p-Tolyl)piperazine dihydrochloride, 97%

$92.97 - $343.54

Chemical Identifiers

Chemical Name or Material 1-(p-Tolyl)piperazine dihydrochloride
CAS 13078-14-3
Molecular Formula C11H16N2·2HCl
Formula Weight 249.19
MDL Number MFCD00035286
Beilstein 5639546
Synonym N-(4-Methylphenyl)piperazine dihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0543206

  SDS
alfa aesar™
L0543206
5g Each for $92.97

AAL0543214

  SDS
alfa aesar™
L0543214
25g Each for $343.54
Description & Specifications

Specifications

Formula Weight 249.19
Chemical Name or Material 1-(p-Tolyl)piperazine dihydrochloride
MDL Number MFCD00035286
CAS 13078-14-3
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Melting Point 214° to 216°C
Synonym N-(4-Methylphenyl)piperazine dihydrochloride
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Beilstein 5639546
Quantity 5g
Molecular Formula C11H16N2·2HCl
Assay 97%
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.