Please login to your online account to display your discounted pricing

1-(p-Tolyl)-1-cyclopentanecarboxylic acid, 98%, ACROS Organics™

$227.16 - $227.16

Chemical Identifiers

Chemical Name or Material 1-(p-Tolyl)-1-cyclopentanecarboxylic acid
CAS 80789-75-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C13 H16 O2
Linear Formula CH3C6H4C5H8CO2H
Formula Weight 204.27
MDL Number MFCD00044672
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC170730050

  SDS
thermo fisher scientific
170730050
Glass bottle 5g Each for $227.16
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Powder and Granules
Formula Weight 204.27
Quantity 5g
Color Beige-gray
Melting Point 181° to 184°C
CAS Max % 100.0
CAS 80789-75-9
Assay Percent Range 98%
Chemical Name or Material 1-(p-Tolyl)-1-cyclopentanecarboxylic acid
CAS Min % 97.5
MDL Number MFCD00044672
Molecular Formula C13 H16 O2
Linear Formula CH3C6H4C5H8CO2H
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Conditions assay on the dry substance
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.