Please login to your online account to display your discounted pricing

1-(p-Tolyl)-1-cyclopentanecarboxylic acid, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  170730050

 View more versions of this product

Catalog No. AC170730050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Powder and Granules
Formula Weight 204.27
Quantity 5g
Color Beige-gray
Melting Point 181° to 184°C
CAS Max % 100.0
Chemical Name or Material 1-(p-Tolyl)-1-cyclopentanecarboxylic acid
Assay Percent Range 98%
CAS 80789-75-9
Infrared Spectrum Authentic
Molecular Formula C13 H16 O2
Linear Formula CH3C6H4C5H8CO2H
Packaging Glass bottle
MDL Number MFCD00044672
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Conditions assay on the dry substance

Specifications

Chemical Name or Material 1-(p-Tolyl)-1-cyclopentanecarboxylic acid
CAS 80789-75-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C13 H16 O2
Linear Formula CH3C6H4C5H8CO2H
Formula Weight 204.27
MDL Number MFCD00044672
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents