missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-(p-Tolyl)-1-cyclohexanecarboxylic Acid 98.0+%, TCI America™
Supplier: TCI America T309725G
Specifications
1-(p-Tolyl)-1-cyclohexanecarboxylic Acid | |
171°C | |
C14H18O2 | |
25 g | |
BEVYUARQHKRBMH-UHFFFAOYSA-N | |
1-(4-methylphenyl)cyclohexane-1-carboxylic acid | |
98664 | |
≥98.0% (GC,T) |
84682-27-9 | |
White-Yellow | |
MFCD00065264 | |
1-4-methylphenyl cyclohexanecarboxylic acid, 1-4-methylphenyl-1-cyclohexanecarboxylic acid, 1-p-tolyl cyclohexanecarboxylic acid, 1-p-tolyl-cyclohexanecarboxylic acid, 1-4-methylphenyl cyclohexane-1-carboxylic acid, 1-p-tolyl-1-cyclohexanecarboxylic acid, 1-p-tolylcyclohexanecarboxylic acid, 1-p-tolyl-1-cyclohexanecarboxylicacid, cyclohexanecarboxylicacid, 1-4-methylphenyl, cyclohexanecarboxylic acid, 1-4-methylphenyl | |
CC1=CC=C(C=C1)C1(CCCCC1)C(O)=O | |
218.30 | |
218.30 | |
Crystalline Powder |
Chemical Identifiers
84682-27-9 | |
218.30 | |
BEVYUARQHKRBMH-UHFFFAOYSA-N | |
98664 | |
CC1=CC=C(C=C1)C1(CCCCC1)C(O)=O |
C14H18O2 | |
MFCD00065264 | |
1-4-methylphenyl cyclohexanecarboxylic acid, 1-4-methylphenyl-1-cyclohexanecarboxylic acid, 1-p-tolyl cyclohexanecarboxylic acid, 1-p-tolyl-cyclohexanecarboxylic acid, 1-4-methylphenyl cyclohexane-1-carboxylic acid, 1-p-tolyl-1-cyclohexanecarboxylic acid, 1-p-tolylcyclohexanecarboxylic acid, 1-p-tolyl-1-cyclohexanecarboxylicacid, cyclohexanecarboxylicacid, 1-4-methylphenyl, cyclohexanecarboxylic acid, 1-4-methylphenyl | |
1-(4-methylphenyl)cyclohexane-1-carboxylic acid |
Safety and Handling
TSCA : No