Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(p-Toluenesulfonyl)pyrrole, 98%

$51.23 - $159.72

Chemical Identifiers

Chemical Name or Material 1-(p-Toluenesulfonyl)pyrrole
CAS 17639-64-4
Molecular Formula C11H11NO2S
Formula Weight 221.28
MDL Number MFCD00145014
Synonym N-Tosylpyrrole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2864106

  SDS
alfa aesar™
H2864106
5g Each for $51.23

AAH2864114

  SDS
alfa aesar™
H2864114
25g Each for $159.72
Description & Specifications

Specifications

Formula Weight 221.28
Synonym N-Tosylpyrrole
Melting Point 99° to 103°C
CAS 17639-64-4
Quantity 5g
Molecular Formula C11H11NO2S
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00145014
Health Hazard 1 Warning
Assay 98%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 1-(p-Toluenesulfonyl)pyrrole
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.