missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(p-Toluenesulfonyl)imidazole, TCI America™

Manufacturer:  TCI America T198525G

 View more versions of this product

Catalog No. T198525G



Specifications

Specifications

1-(p-Toluenesulfonyl)imidazole
77°C
2232-08-8
MFCD00005285
YJYMYJRAQYREBT-UHFFFAOYSA-N
1-(4-methylphenyl)sulfonylimidazole
75219
Crystal-Powder at 20°C
White-Yellow
25g
C10H10N2O2S
1-p-toluenesulfonyl imidazole, 1-tosyl-1h-imidazole, n-tosylimidazole, 1-4-methylphenyl sulfonyl-1h-imidazole, 1-tosylimidazole, 1-p-toluenesulphonyl imidazole, 1-p-tolylsulfonyl imidazole, 1h-imidazole, 1-4-methylphenyl sulfonyl, 1-toluene-p-sulphonyl imidazole, 1-4-methylbenzenesulfonyl imidazole
CC1=CC=C(C=C1)S(=O)(=O)N2C=CN=C2
222.26
222.26
≥98.0% (HPLC,N)

Chemical Identifiers

2232-08-8
222.26
YJYMYJRAQYREBT-UHFFFAOYSA-N
75219
CC1=CC=C(C=C1)S(=O)(=O)N2C=CN=C2
C10H10N2O2S
MFCD00005285
1-p-toluenesulfonyl imidazole, 1-tosyl-1h-imidazole, n-tosylimidazole, 1-4-methylphenyl sulfonyl-1h-imidazole, 1-tosylimidazole, 1-p-toluenesulphonyl imidazole, 1-p-tolylsulfonyl imidazole, 1h-imidazole, 1-4-methylphenyl sulfonyl, 1-toluene-p-sulphonyl imidazole, 1-4-methylbenzenesulfonyl imidazole
1-(4-methylphenyl)sulfonylimidazole
Safety and Handling

Safety and Handling

TSCA : No

Documents