missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(p-Toluenesulfonyl)imidazole, 98+%

Manufacturer:  Alfa Aesar™ H5419122

 View more versions of this product

Catalog No. AAH5419122



Specifications

Specifications

1-(p-Toluenesulfonyl)imidazole
76°C to 78°C
2232-08-8
MFCD00005285
1-4-methylbenzenesulfonyl imidazole
CC1=CC=C(C=C1)S(=O)(=O)N2C=CN=C2
222.26
222.26
Moisture sensitive
100g
C10H10N2O2S
612451
YJYMYJRAQYREBT-UHFFFAOYSA-N
1-(4-methylphenyl)sulfonylimidazole
75219
≥98%

Chemical Identifiers

2232-08-8
222.26
YJYMYJRAQYREBT-UHFFFAOYSA-N
75219
CC1=CC=C(C=C1)S(=O)(=O)N2C=CN=C2
C10H10N2O2S
MFCD00005285
1-4-methylbenzenesulfonyl imidazole
1-(4-methylphenyl)sulfonylimidazole
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 218-771-9

TSCA : No

Recommended Storage : Ambient temperatures

Documents