missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(p-Toluenesulfonyl)-3-nitro-1,2,4-triazole, TCI America™

Manufacturer:  TCI America T29305G

 View more versions of this product

Catalog No. T29305G



Specifications

Specifications

1-(p-Toluenesulfonyl)-3-nitro-1,2,4-triazole
134°C
77451-51-5
MFCD00009756
ZQMJAWSQRGYFBM-UHFFFAOYSA-N
1-(4-methylphenyl)sulfonyl-3-nitro-1,2,4-triazole
688072
Crystal-Powder at 20°C
White-Yellow
5g
C9H8N4O4S
3-nitro-1-tosyl-1h-1,2,4-triazole, 1-p-toluenesulfonyl-3-nitro-1,2,4-triazole, 1-4-toluenesulfonyl-3-nitro-1,2,4-triazole, 1-4-methylphenyl sulfonyl-3-nitro-1,2,4-triazole, 1-4-methylbenzenesulfonyl-3-nitro-1,2,4-triazole, tsnt, 3-nitro-1-tosyl-1,2,4-triazole, 1-p-toluenesulfonyl-3-nitro-1,2,4-triazol, 3-nitro-1-p-tolylsulfonyl-1,2,4-triazole, 3-nitro-1-p-toluenesulfonyl-1,2,4-triazole
CC1=CC=C(C=C1)S(=O)(=O)N2C=NC(=N2)[N+](=O)[O-]
268.247
268.25
≥98.0% (HPLC)

Chemical Identifiers

77451-51-5
268.247
ZQMJAWSQRGYFBM-UHFFFAOYSA-N
688072
CC1=CC=C(C=C1)S(=O)(=O)N2C=NC(=N2)[N+](=O)[O-]
C9H8N4O4S
MFCD00009756
3-nitro-1-tosyl-1h-1,2,4-triazole, 1-p-toluenesulfonyl-3-nitro-1,2,4-triazole, 1-4-toluenesulfonyl-3-nitro-1,2,4-triazole, 1-4-methylphenyl sulfonyl-3-nitro-1,2,4-triazole, 1-4-methylbenzenesulfonyl-3-nitro-1,2,4-triazole, tsnt, 3-nitro-1-tosyl-1,2,4-triazole, 1-p-toluenesulfonyl-3-nitro-1,2,4-triazol, 3-nitro-1-p-tolylsulfonyl-1,2,4-triazole, 3-nitro-1-p-toluenesulfonyl-1,2,4-triazole
1-(4-methylphenyl)sulfonyl-3-nitro-1,2,4-triazole
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer

Documents