Please login to your online account to display your discounted pricing

1-(p-Toluenesulfonyl)-3-nitro-1,2,4-triazole, 98%, ACROS Organics™


Quantity 1g

Manufacturer: thermo fisher scientific  AC223250010

 View more versions of this product

Catalog No. AC223250010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder or Crystalline Powder
Formula Weight 268.25
Quantity 1g
Color White to yellow
Assay Percent Range 98%
CAS 77451-51-5
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 1-(p-Toluenesulfonyl)-3-nitro-1, 2, 4-triazole
Molecular Formula C9H8N4O4S
Elemental Analysis 39.29 to 41.30% C, 20.36 to 21.41% N
CAS Min % 97.5
CAS Max % 100.0
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 1-(p-Toluenesulfonyl)-3-nitro-1, 2, 4-triazole
CAS 77451-51-5
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C9H8N4O4S
Formula Weight 268.25
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents