Please login to your online account to display your discounted pricing

1-(p-Toluenesulfonyl)-3-nitro-1,2,4-triazole, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1-(p-Toluenesulfonyl)-3-nitro-1, 2, 4-triazole
CAS 77451-51-5
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C9H8N4O4S
Formula Weight 268.25
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC223250010

 
thermo fisher scientific
AC223250010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC223250050

 
thermo fisher scientific
AC223250050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 268.25
Physical Form Powder or Crystalline Powder
Quantity 1g
Color White to yellow
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 77451-51-5
Assay Percent Range 98%
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Max % 100.0
Chemical Name or Material 1-(p-Toluenesulfonyl)-3-nitro-1, 2, 4-triazole
CAS Min % 97.5
Molecular Formula C9H8N4O4S
Elemental Analysis 39.29 to 41.30% C, 20.36 to 21.41% N
Infrared Spectrum Authentic
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.