missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(p-Toluenesulfonyl)-1,2,4-triazole, TCI America™

Manufacturer:  TCI America T31875G

 View more versions of this product

Catalog No. T31875G



Specifications

Specifications

1-(p-Toluenesulfonyl)-1,2,4-triazole
107°C
13578-51-3
MFCD00458591
GFWABQNSSIQCLB-UHFFFAOYSA-N
1-(4-methylphenyl)sulfonyl-1,2,4-triazole
3505424
Crystal-Powder at 20°C
White
5g
C9H9N3O2S
1-Tosyl-1,2,4-triazole
CC1=CC=C(C=C1)S(=O)(=O)N2C=NC=N2
223.25
223.25
≥98.0% (HPLC)

Chemical Identifiers

13578-51-3
223.25
GFWABQNSSIQCLB-UHFFFAOYSA-N
3505424
CC1=CC=C(C=C1)S(=O)(=O)N2C=NC=N2
C9H9N3O2S
MFCD00458591
1-Tosyl-1,2,4-triazole
1-(4-methylphenyl)sulfonyl-1,2,4-triazole
Safety and Handling

Safety and Handling

TSCA : No

Documents