missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Octyne 95.0+%, TCI America™

Manufacturer:  TCI America O0050250ML

 View more versions of this product

Catalog No. O0050250ML



Specifications

Specifications

1-Octyne
Yellow
629-05-0
MFCD00009546
1-octyne
CCCCCCC#C
110.2
110.20
≥95.0% (GC)
126°C
250mL
C8H14
3295
UMIPWJGWASORKV-UHFFFAOYSA-N
oct-1-yne
12370
Clear Liquid at 20°C

Chemical Identifiers

629-05-0
110.2
UMIPWJGWASORKV-UHFFFAOYSA-N
12370
CCCCCCC#C
C8H14
MFCD00009546
1-octyne
oct-1-yne
Safety and Handling

Safety and Handling

RTECSNumber : RI2655000

TSCA : Yes

Documents