missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Specifications
1-Octyne | |
Yellow | |
C8H14 | |
100 mL | |
1-octyne, hexylacetylene, octyne-1, n-octyne, hex-1-yl acetylene, pubchem13059, acmc-209na4, 4-01-00-01034 beilstein handbook reference, 1-octyne 10g | |
CCCCCCC#C | |
110.20 | |
110.20 | |
Liquid |
629-05-0 | |
126°C | |
MFCD00009546 | |
3295 | |
UMIPWJGWASORKV-UHFFFAOYSA-N | |
oct-1-yne | |
12370 | |
≥95.0% (GC) |
Chemical Identifiers
629-05-0 | |
110.20 | |
UMIPWJGWASORKV-UHFFFAOYSA-N | |
12370 | |
CCCCCCC#C |
C8H14 | |
MFCD00009546 | |
1-octyne, hexylacetylene, octyne-1, n-octyne, hex-1-yl acetylene, pubchem13059, acmc-209na4, 4-01-00-01034 beilstein handbook reference, 1-octyne 10g | |
oct-1-yne |
Safety and Handling
RTECSNumber : RI2655000
TSCA : Yes