Learn More
1-Octyne, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 223511000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
1-Octyne | |
98.5 | |
0.7400g/mL | |
17°C | |
99% | |
C8H14 | |
CH3(CH2)5C∽CH | |
100mL | |
0.74 | |
Solubility in water: 0.024g/L (25°C) | |
CCCCCCC#C | |
110.20 | |
110.2 |
629-05-0 | |
100.0 | |
127°C to 128°C | |
Authentic | |
Glass bottle | |
1.4150 to 1.417 | |
MFCD00009546 | |
01, 258 | |
1-octyne, hexylacetylene, octyne-1, n-octyne, hex-1-yl acetylene, pubchem13059, acmc-209na4, 4-01-00-01034 beilstein handbook reference, 1-octyne 10g | |
UMIPWJGWASORKV-UHFFFAOYSA-N | |
oct-1-yne | |
12370 | |
99% |
Safety and Handling
GHS H Statement:
May be fatal if swallowed and enters airways.
Harmful if swallowed.
Highly flammable liquid and vapor.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 211-069-3
RUO â Research Use Only