Please login to your online account to display your discounted pricing

1-Octyne, 99%, ACROS Organics™


Quantity 25mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  223510250

 View more versions of this product

Catalog No. AC223510250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Liquid
Formula Weight 110.2
Quantity 25mL
Color Clear colorless to yellow
Beilstein 01, 258
Density 0.74g/mL
CAS Max % 100.0
Chemical Name or Material 1-Octyne
Infrared Spectrum Authentic
Linear Formula CH3(CH2)5C2CH
Packaging Glass bottle
Refractive Index 1.4150 to 1.4170 (20°C, 589nm)
Assay Percent Range 99%
Molecular Formula C8H14
CAS 629-05-0
CAS Min % 98.5
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00009546
Health Hazard 2 GHS H Statement:
May be fatal if swallowed and enters airways.
Harmful if swallowed.
Highly flammable liquid and vapor.

Specifications

Chemical Name or Material 1-Octyne
CAS 629-05-0
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H14
Linear Formula CH3(CH2)5C2CH
Formula Weight 110.2
MDL Number MFCD00009546
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May be fatal if swallowed and enters airways.
Harmful if swallowed.
Highly flammable liquid and vapor.

SDS & Safety Documents