missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Octyne, 98%, Alfa Aesar™

Manufacturer:  Alfa Aesar A1244022

 View more versions of this product

Catalog No. AAA1244022



Description

Description

Applications
1-Octyne is widely used in organic reactions such as halogenation, hydration, oxidative cleavage, hydrogenation, hydrohalogenation, nitrile formation, polymerization and substitution reactions. It is used a precursor for the preparation of monohalogen substituted akenes or dihalogen substituted alkanes on treated with hydrogen halides.

Solubility
Miscible with alcohol, ether and other organic solvents. Immiscible with water.

Notes
Incompatible with oxidizing agents. Keep away from heat, sparks, sources of ignition and flame.
Specifications

Specifications

1-Octyne
100g
98%
CH3(CH2)5C≡CH
UN3295
1-octyne, hexylacetylene, octyne-1, n-octyne, hex-1-yl acetylene, pubchem13059, 1-octyne, acmc-209na4, 4-01-00-01034 beilstein handbook reference, 1-octyne 10g
CCCCCCC#C
110.2
110.2
98%
0.746
629-05-0
C8H14
MFCD00009546
1734494
UMIPWJGWASORKV-UHFFFAOYSA-N
oct-1-yne
12370
Liquid

Chemical Identifiers

629-05-0
110.2
UMIPWJGWASORKV-UHFFFAOYSA-N
12370
CCCCCCC#C
C8H14
MFCD00009546
1-octyne, hexylacetylene, octyne-1, n-octyne, hex-1-yl acetylene, pubchem13059, 1-octyne, acmc-209na4, 4-01-00-01034 beilstein handbook reference, 1-octyne 10g
oct-1-yne
Safety and Handling

Safety and Handling

GHS H Statement
H225-H304

GHS P Statement
P210-P243-P301+P330+P331-P315

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.

RTECSNumber : RI2655000

TSCA : TSCA

SDS Documents