missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Octyn-3-ol 98.0+%, TCI America™

Manufacturer:  TCI America O0196250ML

 View more versions of this product

Catalog No. O0196250ML



Specifications

Specifications

1-Octyn-3-ol
Yellow
818-72-4
MFCD00004588
VUGRNZHKYVHZSN-UHFFFAOYSA-N
oct-1-yn-3-ol
13166
Clear Liquid at 20°C
83°C
250mL
C8H14O
1-octyn-3-ol, 1-octyne-3-ol, oct-1-yne-3-ol, 1-oct-1-yn-3-ol, 3-hydroxyoct-1-yne, acmc-20apfv, acmc-20apkf, rac-1-octyn-3-ol, oct-1 yn-3 ol, pubchem11033
CCCCCC(C#C)O
126.199
126.20
≥98.0% (GC)

Chemical Identifiers

818-72-4
126.199
VUGRNZHKYVHZSN-UHFFFAOYSA-N
13166
CCCCCC(C#C)O
C8H14O
MFCD00004588
1-octyn-3-ol, 1-octyne-3-ol, oct-1-yne-3-ol, 1-oct-1-yn-3-ol, 3-hydroxyoct-1-yne, acmc-20apfv, acmc-20apkf, rac-1-octyn-3-ol, oct-1 yn-3 ol, pubchem11033
oct-1-yn-3-ol
Safety and Handling

Safety and Handling

TSCA : Yes

Documents