missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Octylphosphonic acid, 98%

Manufacturer:  Alfa Aesar™ 2065103

 View more versions of this product

Catalog No. AA2065103



Specifications

Specifications

1-Octylphosphonic acid
98%
MFCD00015841
1-Octanephosphonic acid
CCCCCCCCP(=O)(O)O
194.211
194.21
100°C to 102°C
4724-48-5
C8H19O3P
1764242
NJGCRMAPOWGWMW-UHFFFAOYSA-N
octylphosphonic acid
78452
Crystalline
1g

Chemical Identifiers

4724-48-5
194.211
NJGCRMAPOWGWMW-UHFFFAOYSA-N
78452
CCCCCCCCP(=O)(O)O
C8H19O3P
MFCD00015841
1-Octanephosphonic acid
octylphosphonic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see ... on this label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

TSCA : TSCA

SDS Documents