missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Octanol, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A15977AP

 View more versions of this product

Catalog No. AAA15977AP



Description

Description

1-Octanol is used as a precursor to prepare esters, which is used in perfumes and flavorings. It is used as food additive, anti-foaming agent and emulsifier in anti-rust emulsions. It is widely involved in chemical industry for the synthesis of ethoxylates, alkyl sulfates and ether sulfates. It is used as a solvent in paints, varnishes, waxes, and surface coatings and mainly involved in agricultural chemistry to inhibit excessive growth of tobacco plants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Octanol is used as a precursor to prepare esters, which is used in perfumes and flavorings. It is used as food additive, anti-foaming agent and emulsifier in anti-rust emulsions. It is widely involved in chemical industry for the synthesis of ethoxylates, alkyl sulfates and ether sulfates. It is used as a solvent in paints, varnishes, waxes, and surface coatings and mainly involved in agricultural chemistry to inhibit excessive growth of tobacco plants.

Solubility
Miscible with ethanol, ether and propylene glycol. Immiscible with water.

Notes
Incompatible with strong oxidizing agents. Keep the container tightly closed in a dry and well-ventilated place.
Specifications

Specifications

1-Octanol
°C to 16°C
193°C to 195°C
99%
1.429
MFCD00002988
1697461
1-octanol, octanol, octyl alcohol, n-octanol, capryl alcohol, caprylic alcohol, n-octyl alcohol, heptyl carbinol, 1-hydroxyoctane, primary octyl alcohol
KBPLFHHGFOOTCA-UHFFFAOYSA-N
octan-1-ol
957
130.23
111-87-5
0.825
81°C (177°F)
C8H18O
CH3(CH2)7OH
500 mL
14,6751
Miscible with ethanol,ether and propylene glycol. Immiscible with water.
CCCCCCCCO
130.23
CHEBI:16188
99%

Chemical Identifiers

111-87-5
130.23
KBPLFHHGFOOTCA-UHFFFAOYSA-N
957
octan-1-ol
C8H18O
MFCD00002988
1-octanol, octanol, octyl alcohol, n-octanol, capryl alcohol, caprylic alcohol, n-octyl alcohol, heptyl carbinol, 1-hydroxyoctane, primary octyl alcohol
CHEBI:16188
CCCCCCCCO
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H227

P210-P264b-P280-P305+P351+P338-P370+P378q-P501c

H227-H319

EINECSNumber : 203-917-6

RTECSNumber : RH6550000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only