missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Octanethiol 98.0+%, TCI America™
Supplier: TCI America O0025500ML
Specifications
1-Octanethiol | |
Yellow | |
C8H18S | |
500 mL | |
1-octanethiol, 1-mercaptooctane, n-octyl mercaptan, octyl mercaptan, octanethiol, octylthiol, 1-octylthiol, n-octanethiol, octylmercaptan, n-octylthiol | |
CCCCCCCCS | |
146.29 | |
146.29 | |
Liquid |
111-88-6 | |
200°C | |
MFCD00004912 | |
3082 | |
KZCOBXFFBQJQHH-UHFFFAOYSA-N | |
octane-1-thiol | |
8144 | |
≥98.0% (GC) |
Chemical Identifiers
111-88-6 | |
146.29 | |
KZCOBXFFBQJQHH-UHFFFAOYSA-N | |
8144 | |
CCCCCCCCS |
C8H18S | |
MFCD00004912 | |
1-octanethiol, 1-mercaptooctane, n-octyl mercaptan, octyl mercaptan, octanethiol, octylthiol, 1-octylthiol, n-octanethiol, octylmercaptan, n-octylthiol | |
octane-1-thiol |
Safety and Handling
EINECSNumber : (2)-0464
TSCA : Yes