missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Octanethiol, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 157482500

 View more versions of this product

Catalog No. AC157482500



Specifications

Specifications

1-Octanethiol
68°C
Glass bottle
0.84
Colorless
250g
96.0
96% min. (GC)
CH3(CH2)7SH
1-mercaptooctane
CCCCCCCCS
146.29
146.29
97%
0.8400g/mL
Authentic
1.4525 to 1.4545
197.0°C to 200.0°C
-49.0°C
111-88-6
100.0
C8H18S
MFCD00004912
KZCOBXFFBQJQHH-UHFFFAOYSA-N
octane-1-thiol
8144
Liquid

Chemical Identifiers

111-88-6
146.29
KZCOBXFFBQJQHH-UHFFFAOYSA-N
8144
CCCCCCCCS
C8H18S
MFCD00004912
1-mercaptooctane
octane-1-thiol
Safety and Handling

Safety and Handling

GHS H Statement
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Wear eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

GHS Signal Word: Warning

EINECSNumber : 203-918-1

SDS Documents