Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Octanesulfonyl chloride, 97%

Chemical Identifiers

Chemical Name or Material 1-Octanesulfonyl chloride
CAS 7795-95-1
Molecular Formula C8H17ClO2S
Linear Formula CH3(CH2)7SO2Cl
Formula Weight 212.74
MDL Number MFCD00014729
UN Number UN3265
Beilstein 1763921
Synonym 1-Octylsulfonyl chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1544009

  SDS
alfa aesar™
A1544009
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1544030

  SDS
alfa aesar™
A1544030
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1544018

  SDS
alfa aesar™
A1544018
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 212.74
Chemical Name or Material 1-Octanesulfonyl chloride
TSCA TSCA
Assay 97%
Molecular Formula C8H17ClO2S
Linear Formula CH3(CH2)7SO2Cl
MDL Number MFCD00014729
CAS 7795-95-1
UN Number UN3265
Beilstein 1763921
Synonym 1-Octylsulfonyl chloride
Health Hazard 1 Danger
Boiling Point 75°C (0.03mmHg)
Melting Point 14°C
Quantity 10g
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flash Point >110°C (230°F)
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.