Please login to your online account to display your discounted pricing

1-Octanesulfonic Acid Sodium Salt, HPLC Grade, J.T.Baker™


High purity solvents for use in analytical and preparative separations

Quantity 25g
Packaging Wide Mouth Amber Glass Bottle

Manufacturer: avantor performance  281805

 View more versions of this product

Catalog No. 02002045

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 216.27
Packaging Wide Mouth Amber Glass Bottle
Molecular Formula CH3(CH2)7SO3Na
CAS 5324-84-5
Chemical Name or Material 1-Octanesulfonic Acid Sodium Salt
Purity Grade HPLC Grade
Specific Gravity 1L = 0.00kg
Quantity 25g

Specifications

Chemical Name or Material 1-Octanesulfonic Acid Sodium Salt
CAS 5324-84-5
Purity Grade HPLC Grade
Molecular Formula CH3(CH2)7SO3Na
Formula Weight 216.27

    For Ion-Pair Chromatography of Basic Compounds