Please login to your online account to display your discounted pricing

1-Octanesulfonic Acid Sodium Salt, HPLC Grade, J.T.Baker™

High purity solvents for use in analytical and preparative separations

$256.24 - $3,367.58

Chemical Identifiers

Chemical Name or Material 1-Octanesulfonic Acid Sodium Salt
CAS 5324-84-5
Purity Grade HPLC Grade
Molecular Formula CH3(CH2)7SO3Na
Formula Weight 216.27
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002045

 
avantor performance
281805
Wide Mouth Amber Glass Bottle 25g Each for $256.24

02002046

 
avantor performance
281801
Wide Mouth Amber Glass Bottle 500g Each for $3,367.58
Description & Specifications

Specifications

Formula Weight 216.27
Packaging Wide Mouth Amber Glass Bottle
Molecular Formula CH3(CH2)7SO3Na
CAS 5324-84-5
Chemical Name or Material 1-Octanesulfonic Acid Sodium Salt
Purity Grade HPLC Grade
Specific Gravity 1L = 0.00kg
Quantity 25g

    For Ion-Pair Chromatography of Basic Compounds