Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Octadecanol, 97%

$34.04 - $143.15

Chemical Identifiers

Chemical Name or Material 1-Octadecanol
CAS 112-92-5
Molecular Formula CH3(CH2)17OH
Formula Weight 270.5
MDL Number MFCD00002823
Beilstein 1362907
Merck Index 14,8805
Synonym n-Octadecyl alcohol; Stearyl alcohol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1202036

  SDS
alfa aesar
A1202036
500g Each for $39.49

AAA1202030

  SDS
alfa aesar™
A1202030
250g Each for $34.04

AAA120200E

  SDS
alfa aesar™
A120200E
2500g Each for $143.15
Description & Specifications

Specifications

Formula Weight 270.5
Quantity 500g
TSCA TSCA
Assay 97%
MDL Number MFCD00002823
Beilstein 1362907
Merck Index 14,8805
RTECS Number RG2010000
Chemical Name or Material 1-Octadecanol
Assay Percent Range 97%
Molecular Formula CH3(CH2)17OH
CAS 112-92-5

Applications
1-Octadecanol has been used as surfactant for the functionalization of carbon nanotubes. It is employed as emulsion stabilizer for creams and lotions, quality modifier of lipsticks, additive for ointment base and cream conditioners for smooth touch to skin for cosmetic uses. It is auxiliary for emulsifiers.

Solubility
Insoluble in water. Soluble in alcohol, acetone, ether, benzene, chloroform.

Notes
Store away from oxidizing agents. Keep the container tightly closed in a well ventilated place.