missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Octadecanol, 97%, Alfa Aesar™

Manufacturer:  Alfa Aesar A1202030

 View more versions of this product

Catalog No. AAA1202030



Description

Description

Applications
1-Octadecanol has been used as surfactant for the functionalization of carbon nanotubes. It is employed as emulsion stabilizer for creams and lotions, quality modifier of lipsticks, additive for ointment base and cream conditioners for smooth touch to skin for cosmetic uses. It is auxiliary for emulsifiers.

Solubility
Insoluble in water. Soluble in alcohol, acetone, ether, benzene, chloroform.

Notes
Store away from oxidizing agents. Keep the container tightly closed in a well ventilated place.
Specifications

Specifications

1-Octadecanol
210°C (15mmHg)
250g
97%
MFCD00002823
14,8805
GLDOVTGHNKAZLK-UHFFFAOYSA-N
octadecan-1-ol
8221
270.5
174°C (345°F)
56°C to 60°C
112-92-5
C18H38O
1362907
stearyl alcohol, 1-octadecanol, octadecanol, octadecyl alcohol, n-octadecanol, 1-hydroxyoctadecane, n-1-octadecanol, stearol, n-octadecyl alcohol, stearic alcohol
CCCCCCCCCCCCCCCCCCO
270.501
CHEBI:32154
97%

Chemical Identifiers

112-92-5
270.501
GLDOVTGHNKAZLK-UHFFFAOYSA-N
8221
octadecan-1-ol
C18H38O
MFCD00002823
stearyl alcohol, 1-octadecanol, octadecanol, octadecyl alcohol, n-octadecanol, 1-hydroxyoctadecane, n-1-octadecanol, stearol, n-octadecyl alcohol, stearic alcohol
CHEBI:32154
CCCCCCCCCCCCCCCCCCO
Safety and Handling

Safety and Handling

RTECSNumber : RG2010000

TSCA : TSCA

SDS Documents