missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Octacosanol, 90+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC258130010

 View more versions of this product

Catalog No. AC258130010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

1-Octacosanol
Glass Bottle
White
1g
90.0
90% min. (GC)
CH3(CH2)27OH
15, 6833
Solubility in water: insoluble. Other solubilities: soluble in hot hydrocarbons, hot dibutylether,, thf,,hot methylisobutylketone
CCCCCCCCCCCCCCCCCCCCCCCCCCCCO
410.77
CHEBI:28243
Crystalline Powder or Flaked Crystals
Authentic
200.0°C to 250.0°C
83.0°C
557-61-9
100.0
C28H58O
MFCD00044770
1-octacosanol, cluytyl alcohol, montanyl alcohol, n-octacosanol, octacosanol, octacosanol-1, octacosyl, octacosyl alcohol, policosanol, polycosanol
CNNRPFQICPFDPO-UHFFFAOYSA-N
octacosan-1-ol
68406
410.77
90+%

Chemical Identifiers

557-61-9
410.77
CNNRPFQICPFDPO-UHFFFAOYSA-N
68406
octacosan-1-ol
C28H58O
MFCD00044770
1-octacosanol, cluytyl alcohol, montanyl alcohol, n-octacosanol, octacosanol, octacosanol-1, octacosyl, octacosyl alcohol, policosanol, polycosanol
CHEBI:28243
CCCCCCCCCCCCCCCCCCCCCCCCCCCCO
Safety and Handling

Safety and Handling

EINECSNumber : 209-181-2

SDS Documents