missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(o-Tolyl)piperazine dihydrochloride, 98%

$65.92 - $667.12

Chemical Identifiers

CAS 70849-60-4
Molecular Formula C11H17ClN2
Molecular Weight (g/mol) 212.721
MDL Number MFCD00039030
InChI Key LZNYIWJDVYCKDU-UHFFFAOYSA-N
Synonym 1-2-methylphenyl piperazine hydrochloride, 1-2-tolyl piperazine hydrochloride, 1-o-tolyl piperazine hydrochloride, 1-o-tolyl piperazinehydrochloride, 1-o-tolyl-piperazine hydrochloride, acmc-209ryj, n-2-methylphenyl piperazine hydrochloride, n-o-tolyl piperazine hydrochloride, piperazine, 1-2-methylphenyl-, hydrochloride, piperazine, 1-2-methylphenyl-, monohydrochloride
PubChem CID 12227895
IUPAC Name 1-(2-methylphenyl)piperazine;hydrochloride
SMILES CC1=CC=CC=C1N2CCNCC2.Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2260406
View Documents
Alfa Aesar™
B2260406
5g
Each for $65.92
 
AAB2260414
View Documents
Alfa Aesar™
B2260414
25g
Each for $196.96
 
AAB2260422
View Documents
Alfa Aesar™
B2260422
100g
Each for $667.12
 
Description

Description

Specifications

Chemical Identifiers

70849-60-4
212.721
LZNYIWJDVYCKDU-UHFFFAOYSA-N
12227895
CC1=CC=CC=C1N2CCNCC2.Cl
C11H17ClN2
MFCD00039030
1-2-methylphenyl piperazine hydrochloride, 1-2-tolyl piperazine hydrochloride, 1-o-tolyl piperazine hydrochloride, 1-o-tolyl piperazinehydrochloride, 1-o-tolyl-piperazine hydrochloride, acmc-209ryj, n-2-methylphenyl piperazine hydrochloride, n-o-tolyl piperazine hydrochloride, piperazine, 1-2-methylphenyl-, hydrochloride, piperazine, 1-2-methylphenyl-, monohydrochloride
1-(2-methylphenyl)piperazine;hydrochloride

Specifications

Hygroscopic
5g
C11H17ClN2
4019516
LZNYIWJDVYCKDU-UHFFFAOYSA-N
1-(2-methylphenyl)piperazine;hydrochloride
12227895
98%
238°C to 240°C
70849-60-4
MFCD00039030
1-2-methylphenyl piperazine hydrochloride, 1-2-tolyl piperazine hydrochloride, 1-o-tolyl piperazine hydrochloride, 1-o-tolyl piperazinehydrochloride, 1-o-tolyl-piperazine hydrochloride, acmc-209ryj, n-2-methylphenyl piperazine hydrochloride, n-o-tolyl piperazine hydrochloride, piperazine, 1-2-methylphenyl-, hydrochloride, piperazine, 1-2-methylphenyl-, monohydrochloride
CC1=CC=CC=C1N2CCNCC2.Cl
212.721
249.19
1-(o-Tolyl)piperazine dihydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c

H302-H311-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: II

TSCA : No

Recommended Storage : Ambient temperatures

SDS