Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-O-Palmitoyl-2-O-acetyl-sn-glycero-3-phosphocholine, 99%


Quantity 5mg

Manufacturer: alfa aesar™  J67083LB0

 View more versions of this product

Catalog No. AAJ67083LB0

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 537.67
Chemical Name or Material 1-O-Palmitoyl-2-O-acetyl-sn-glycero-3-phosphocholine
CAS 84062-61-3
MDL Number MFCD00798242
Physical Form Viscous Oil
Assay 99%
Storage Note 1 -30° to -10°C
Quantity 5mg
Synonym 1-Acyl-PAF

Specifications

Chemical Name or Material 1-O-Palmitoyl-2-O-acetyl-sn-glycero-3-phosphocholine
CAS 84062-61-3
Formula Weight 537.67
MDL Number MFCD00798242
Synonym 1-Acyl-PAF
Accessories

-30° to -10°C

SDS & Safety Documents