Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-O-Palmitoyl-2-O-acetyl-sn-glycero-3-phosphocholine, 99%

Chemical Identifiers

Chemical Name or Material 1-O-Palmitoyl-2-O-acetyl-sn-glycero-3-phosphocholine
CAS 84062-61-3
Formula Weight 537.67
MDL Number MFCD00798242
Synonym 1-Acyl-PAF
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67083LB0

  SDS
alfa aesar™
J67083LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67083MCR

  SDS
alfa aesar™
J67083MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 537.67
Chemical Name or Material 1-O-Palmitoyl-2-O-acetyl-sn-glycero-3-phosphocholine
CAS 84062-61-3
MDL Number MFCD00798242
Physical Form Viscous Oil
Assay 99%
Storage Note 1 -30° to -10°C
Quantity 5mg
Synonym 1-Acyl-PAF
Safety and Handling

-30° to -10°C