missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-O-n-Octyl-β-D-glucopyranoside, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 230342500
Specifications
(5% aq. soln.) clear, colorless | |
Authentic | |
− 32.00 (20.00°C c=5,water) | |
White | |
250mg | |
97.5 | |
97.5% min. (ELSD) (HPLC) | |
MFCD00063288 | |
2r,3s,4s,5r,6r-2-hydroxymethyl-6-octyloxy tetrahydro-2h-pyran-3,4,5-triol, b-octylglucoside, n-octyl glucoside, n-octyl-beta-d-glucoside, octyl b-d-glucopyranoside, octyl beta-d-glucopyranoside, octyl beta-d-glucoside, octyl glucoside, octyl-beta-d-glucopyranoside, unii-v109wut6rl | |
HEGSGKPQLMEBJL-RKQHYHRCSA-N | |
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-octoxyoxane-3,4,5-triol | |
62852 | |
Crystalline Powder and Chunks |
1-O-n-Octyl-β-D-glucopyranoside | |
Glass bottle | |
-32 | |
105°C | |
29836-26-8 | |
100.0 | |
C14H28O6 | |
15, 6857 | |
(10% in water) Clear to slightly hazy colorless to light yellow | |
CCCCCCCCOC1C(C(C(C(O1)CO)O)O)O | |
292.36 | |
292.36 | |
98% |
Chemical Identifiers
29836-26-8 | |
292.36 | |
HEGSGKPQLMEBJL-RKQHYHRCSA-N | |
62852 | |
CCCCCCCCOC1C(C(C(C(O1)CO)O)O)O |
C14H28O6 | |
MFCD00063288 | |
2r,3s,4s,5r,6r-2-hydroxymethyl-6-octyloxy tetrahydro-2h-pyran-3,4,5-triol, b-octylglucoside, n-octyl glucoside, n-octyl-beta-d-glucoside, octyl b-d-glucopyranoside, octyl beta-d-glucopyranoside, octyl beta-d-glucoside, octyl glucoside, octyl-beta-d-glucopyranoside, unii-v109wut6rl | |
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-octoxyoxane-3,4,5-triol |
Safety and Handling
EINECSNumber : 249-887-8